Learn More
Thermo Scientific™ Smart-Vue™ Client Server Software
Catalog No. SV612500LSB
Change view
Click to view available options
Sensor Types:
20 Sensors
20 to 49 Sensors
20 to 99 Sensors
20 to Unlimited Sensors
49 Sensors
49 to 99 Sensors
49 to Unlimited Sensors
99 Sensors
99 to Unlimited Sensors
Unlimited Sensors
Software Type:
Smart-Vue Software 2.0
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sensor Types Software Type
SV612500LSB 99 to Unlimited Sensors Smart-Vue Software 2.0
SV603500LSB 20 Sensors Smart-Vue Software 2.0
SV604500LSB 49 Sensors Smart-Vue Software 2.0
SV605500LSB 99 Sensors Smart-Vue Software 2.0
SV606500LSB Unlimited Sensors Smart-Vue Software 2.0
SV607500LSB 20 to 49 Sensors Smart-Vue Software 2.0
SV608500LSB 20 to 99 Sensors Smart-Vue Software 2.0
SV609500LSB 20 to Unlimited Sensors Smart-Vue Software 2.0
SV610500LSB 49 to 99 Sensors Smart-Vue Software 2.0
SV611500LSB 49 to Unlimited Sensors Smart-Vue Software 2.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. SV612500LSB Supplier Thermo Scientific™ Supplier No. SV612500LSB
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Automate monitoring of vital laboratory equipment parameters with Thermo Scientific™ Smart-Vue™ Client Server Software and enjoy greater peace of mind.
Automate monitoring of vital laboratory equipment parameters with Thermo Scientific™ Smart-Vue™ Client Server Software. Find peace of mind knowing your work is fully monitored 24/7 with immediate notifications reaching you even when you are away from the lab. This software provides a plug and play solution with an intuitive interface.
Intelligent Plug & Play Simplicity
  • Installation Wizard for simplified, step-by-step process
  • Service Discovery Protocol (SDP) enables every module to send signal to receiver—identifies most efficient communication route based on RF signal strength
  • Module automatically establishes new path to receiver if no communication during 6-hour period
  • Back-up system with memory capacity up to 3000 points, with uninterruptible power for continuous data logging
Intuitive Dashboard
  • Daily usage is simplified
  • Single-click export of reports to Microsoft™ Excel™, Word™ or Adobe™ PDF formats
  • Flexible creation of graphs summarizing data from individual or multiple sensors
  • Visual depiction of RF signal strength an QoS as monitored by each module
Data Traceability & Security
  • Secure, customized levels of user access
  • Audit trails
  • Archived alarms
  • Password protection
  • Secure digital transmission and data back-ups
Numerous Configuration Capabilities
  • Combines RF and TCP/IP network technologies
  • Share equipment data over entire network instantly
  • Download simple plug-in to visualize equipment data readings from any workstation
Compatible with:
Refrigerators, Freezers, LN2 Storage, CO2 Incubators, Clean Rooms, Storage Areas, Ovens
Recommended for:
Ideal for use in highly controlled environments, including pharmaceutical production, biotechnology, IVF clinics, clinical studies and diagnostic laboratories.
Ordering Information:
Availability of Smart-Vue products vary by country. Please ask your local sales representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.
Specifications
Type Sensor Software
Software Type Smart-Vue Software 2.0
Includes User Manuals, Quick Installation Guides, Software Drivers
Certifications/Compliance Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Sensor Types 99 to Unlimited Sensors
For Use With (Equipment) Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven
Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.