Learn More
Thermo Scientific™ Smart-Vue™ Client Server Software
Automate monitoring of vital laboratory equipment parameters with Thermo Scientific™ Smart-Vue™ Client Server Software and enjoy greater peace of mind.
Supplier: Thermo Scientific™ SV610500LSB
Description
Automate monitoring of vital laboratory equipment parameters with Thermo Scientific™ Smart-Vue™ Client Server Software. Find peace of mind knowing your work is fully monitored 24/7 with immediate notifications reaching you even when you are away from the lab. This software provides a plug and play solution with an intuitive interface.
Intelligent Plug & Play Simplicity
- Installation Wizard for simplified, step-by-step process
- Service Discovery Protocol (SDP) enables every module to send signal to receiver—identifies most efficient communication route based on RF signal strength
- Module automatically establishes new path to receiver if no communication during 6-hour period
- Back-up system with memory capacity up to 3000 points, with uninterruptible power for continuous data logging
Intuitive Dashboard
- Daily usage is simplified
- Single-click export of reports to Microsoft™ Excel™, Word™ or Adobe™ PDF formats
- Flexible creation of graphs summarizing data from individual or multiple sensors
- Visual depiction of RF signal strength an QoS as monitored by each module
Data Traceability & Security
- Secure, customized levels of user access
- Audit trails
- Archived alarms
- Password protection
- Secure digital transmission and data back-ups
Numerous Configuration Capabilities
- Combines RF and TCP/IP network technologies
- Share equipment data over entire network instantly
- Download simple plug-in to visualize equipment data readings from any workstation
Compatible with:
Refrigerators, Freezers, LN2 Storage, CO2 Incubators, Clean Rooms, Storage Areas, Ovens
Recommended for:
Ideal for use in highly controlled environments, including pharmaceutical production, biotechnology, IVF clinics, clinical studies and diagnostic laboratories.
Ordering Information:
Availability of Smart-Vue products vary by country. Please ask your local sales representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.
Chemical Identifiers
| Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance. |
Specifications
| Sensor Software | |
| User Manuals, Quick Installation Guides, Software Drivers | |
| 49 to 99 Sensors |
| Smart-Vue Software 2.0 | |
| Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance. | |
| Refrigerator, Freezer, LN2 Storage, CO2 Incubator, Oven |
Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.