Learn More
Leap Technologies Smart Syringe 10Ul 23S
Catalog No. 501920838
Encompass
10uL Smart Syringe with fixed needle for Tool D7/57: Needle length 57mm metal plunger gauge 23S scale length 54mm Point Style conical (Instrument: Pal3)
Catalog No. 501920838 Supplier Leap Technologies Supplier No. SF1057M23SCO
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10uL Smart Syringe with fixed needle for Tool D7/57: Needle length 57mm metal plunger gauge 23S scale length 54mm Point Style conical (Instrument: Pal3)