missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Smart SPME Fiber Assembly, Polydimethylsiloxane (PDMS)

Catalog No. 111023067
Change view
Click to view available options
Color:
Green
Red
Yellow
Thickness (Metric):
7 μm
30 μm
100 μm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Thickness (Metric)
111023067 Red 100 μm
111023066 Yellow 30 μm
111023065 Green 7 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111023067 Supplier MilliporeSigma™ Supelco™ Supplier No. 548575U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Smart SPME fiber assemblies combine innovative SPME devices with Smart technology for seamless sample preparation.

  • Smart SPME fiber assemblies combine innovative SPME devices with Smart technology for seamless sample preparation.
  • Each Smart fiber assembly, specifically designed to be compatible with the PAL 3, Series II Smart autosampler, incorporates a SPME fiber within a holder and a programmable computer chip coded with fiber chemistry type, dimensions, and lot number.
  • The Smart chip provides traceability by recording fiber stroke count, injection and conditioning durations, usage dates, and expiration date.
  • The one piece fiber/holder design makes the Smart SPME assembly easier to handle and install than traditional SPME.
  • In addition, compatibility with the SPME tool on the PAL 3 autosampler allows for more efficient method development and completely automated sample preparation, thus increasing overall productivity.

Specifications

For Use With (Equipment) PAL 3, Series II Smart Autosampler
Type Fiber Assembly
Matrix Polydimethylsiloxane (PDMS)
Needle Gauge 23 G
Thickness (Metric) 100 μm
Color Red