Learn More
MilliporeSigma™ Supelco™ Smart SPME Fiber Assembly, Polydimethylsiloxane (PDMS)
Smart SPME fiber assemblies combine innovative SPME devices with Smart technology for seamless sample preparation.
Supplier: MilliporeSigma™ Supelco™ 548552U
Description
- Smart SPME fiber assemblies combine innovative SPME devices with Smart technology for seamless sample preparation.
- Each Smart fiber assembly, specifically designed to be compatible with the PAL 3, Series II Smart autosampler, incorporates a SPME fiber within a holder and a programmable computer chip coded with fiber chemistry type, dimensions, and lot number.
- The Smart chip provides traceability by recording fiber stroke count, injection and conditioning durations, usage dates, and expiration date.
- The one piece fiber/holder design makes the Smart SPME assembly easier to handle and install than traditional SPME.
- In addition, compatibility with the SPME tool on the PAL 3 autosampler allows for more efficient method development and completely automated sample preparation, thus increasing overall productivity.
Chemical Identifiers
| PAL 3, Series II Smart Autosampler | |
| Fiber Assembly | |
| Polydimethylsiloxane (PDMS) | |
| 23 G |
Specifications
| PAL 3, Series II Smart Autosampler | |
| Polydimethylsiloxane (PDMS) | |
| 7 μm |
| Fiber Assembly | |
| 23 G | |
| Green |