missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Smart SPME Fiber Assembly, Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)
Description
- Smart SPME fiber assemblies combine innovative SPME devices with Smart technology for seamless sample preparation.
- Each Smart fiber assembly, specifically designed to be compatible with the PAL 3, Series II Smart autosampler, incorporates a SPME fiber within a holder and a programmable computer chip coded with fiber chemistry type, dimensions, and lot number.
- The Smart chip provides traceability by recording fiber stroke count, injection and conditioning durations, usage dates, and expiration date.
- The one piece fiber/holder design makes the Smart SPME assembly easier to handle and install than traditional SPME.
- In addition, compatibility with the SPME tool on the PAL 3 autosampler allows for more efficient method development and completely automated sample preparation, thus increasing overall productivity.
Specifications
Specifications
| For Use With (Equipment) | PAL 3, Series II Smart Autosampler |
| Type | Fiber Assembly |
| Matrix | Polydimethylsiloxane/Divinylbenzene (PDMS/DVB) |
| Needle Gauge | 23 G |
| Thickness (Metric) | 65 μm |
| Color | Blue |