missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Smart SPME Fiber Assembly, Carboxen/Polydimethylsiloxane (CAR/PDMS)

Catalog No. p-7226310
Change view
Click to view available options
Color:
Blue
Thickness (Metric):
85 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Thickness (Metric)
111023064 Blue 85 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023064 Supplier MilliporeSigma™ Supelco™ Supplier No. 548551U
Only null left

Smart SPME fiber assemblies combine innovative SPME devices with Smart technology for seamless sample preparation.

  • Smart SPME fiber assemblies combine innovative SPME devices with Smart technology for seamless sample preparation.
  • Each Smart fiber assembly, specifically designed to be compatible with the PAL 3, Series II Smart autosampler, incorporates a SPME fiber within a holder and a programmable computer chip coded with fiber chemistry type, dimensions, and lot number.
  • The Smart chip provides traceability by recording fiber stroke count, injection and conditioning durations, usage dates, and expiration date.
  • The one piece fiber/holder design makes the Smart SPME assembly easier to handle and install than traditional SPME.
  • In addition, compatibility with the SPME tool on the PAL 3 autosampler allows for more efficient method development and completely automated sample preparation, thus increasing overall productivity.

Specifications

For Use With (Equipment) PAL 3, Series 2 Smart Autosampler
Type Fiber Assembly
Material StableFlex Fiber
Matrix Carboxen/Polydimethylsiloxane
Needle Gauge 23 G
Thickness (Metric) 85 μm
Color Blue