missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Smart SPME Fiber Assembly, Carbowax-Polyethylene Glycol (PEG)

Catalog No. 111023073
Change view
Click to view available options
For Use With (Equipment):
PAL 3, Series II Smart Autosampler
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
111023073 PAL 3, Series II Smart Autosampler
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023073 Supplier MilliporeSigma™ Supelco™ Supplier No. 548676U

Smart SPME fiber assemblies combine innovative SPME devices with Smart technology for seamless sample preparation.

  • PAL Smart SPME fiber assemblies combine innovative SPME devices with Smart technology for seamless sample preparation.
  • Each Smart fiber assembly, specifically designed to be compatible with the PAL 3, Series II Smart autosampler, incorporates a SPME fiber within a holder and a programmable computer chip coded with fiber chemistry type, dimensions, and lot number.
  • The Smart chip provides traceability by recording fiber stroke count, injection and conditioning durations, usage dates, and expiration date.
  • The one piece fiber/holder design makes the Smart SPME assembly easier to handle and install than traditional SPME.
  • In addition, compatibility with the SPME tool on the PAL 3 autosampler allows for more efficient method development and completely automated sample preparation, thus increasing overall productivity.

Specifications

For Use With (Equipment) PAL 3, Series II Smart Autosampler
Type Fiber Assembly
Matrix Carbowax-Polyethylene Glycol (PEG)
Needle Gauge 23 G
Thickness (Metric) 60 μm
Color Purple