missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SMART Digest™ RNase Kits

Catalog No. 01257521
Change view
Click to view available options
Enzyme:
RNase A
RNase T1
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Enzyme
01257521 RNase T1
01257522 RNase A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01257521 Supplier Thermo Scientific™ Supplier No. 60120101

Use SMART Digest™ RNase Kits in your oligonucleotide mapping and quantitation workflows to obtain high quality analytical results.

Obtain high-quality analytical results from mRNA digestions using Thermo Scientific™ SMART Digest™ RNase Kits. A significant advance in sample preparation for oligonucleotide mapping and quantitation, these kits provide fast and simple mRNA digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that is compatible with automation.

These RNase kits use optimized, heat stable enzymes on magnetic beads, allowing for control of partial digestion to produce required fragments of ideal length for sequencing by HRAM-MS.

Easy and Simple

  • A simple process and control of digestion time ensure required fragment length for comprehensive mRNA sequencing by HRAM-MS
  • Easily automate the entire protocol for high-throughput processing
  • Single nuclease digestion allows full characterization of mRNA

Reliable Results

  • Increased reproducibility over existing protocols
  • Higher data confidence
  • No nuclease contamination of the analytical column


RNase kits are available with with two endoribonuclease enzyme specificities:

  • RNase T1 degrades single-stranded RNA at G residues
  • RNase A degrades single-stranded RNA at C and U residues

Specifications

Cleavage Degrades single-stranded RNA at G residues
Enzyme RNase T1
Format Bulk Magnetic SMART Digest Resin
For Use With (Application) Oligonucleotide Mapping
Product Line SMART Digest
Type RNA Digestion
Includes Bulk Magnetic SMART Digest Resin, SMART Digest Buffer