missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SMART Digest™ Proteinase K Kits

Catalog No. 13622455
Change view
Click to view available options
Description:
Bulk Resin option
Bulk Resin option with Filter/Collection plate
Bulk Resin option with SOLAμ/collection plate
Magnetic Bulk Resin option
Magnetic Bulk Resin option with Filter/Collection plate
Magnetic Bulk Resin option with SOLAμ/collection plate
with Collection plate
with Filter/Collection plate
with SOLAμ/collection plate
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
13622455 Magnetic Bulk Resin option with Filter/Collection plate
03252212 with Collection plate
03252213 Bulk Resin option
13622454 Magnetic Bulk Resin option
03252214 with Filter/Collection plate
03252215 Bulk Resin option with Filter/Collection plate
03252216 with SOLAμ/collection plate
03252217 Bulk Resin option with SOLAμ/collection plate
03252218 Magnetic Bulk Resin option with SOLAμ/collection plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 13622455 Supplier Thermo Scientific™ Supplier No. 60109108MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well for your biomarker studies.

Obtain high quality analytical results from protein digests by using Thermo Scientific™ SMART Digest™ Proteinase K Kits. A significant advance in sample preparation for biopharmaceutical research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.

Proteinase K Protein Digestion

SMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses proteinase K, which cleaves the peptide bond adjacent to the carboxyl group of aliphatic and aromatic amino acids with blocked alpha amino groups.

Easy and Simple
  • Significantly increased speed of sample processing
  • Simple process
  • Easily automate entire protocol for high-throughput processing
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher sensitivity
  • Higher data confidence
Significantly Reduces Sample Preparation Time

Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.

Cleaner Samples

Reduction in the number of chemicals used and the immobilized proteinase K design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. The results are easier to interpret.

Couple with SOLAμ SPE Plates for Desalting

When used with the SMART Digest for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.

Specifications

Description Magnetic Bulk Resin option with Filter/Collection plate
Capture None
Cleavage Peptide bond adjacent to carboxyl group of aliphatic and aromatic amino acids with blocked alpha amino groups
Enzyme Proteinase K
Format SMART Digest Resin, Bulk Magnetic
For Use With (Application) Peptide Quantitation, Peptide Mapping
Product Line SMART Digest
Type Protein Digestion
Includes Bulk Magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, Filter Plate