Learn More
Greiner Bio-One Small Volume™ 384 Well Solid Bottom Microplates for Compound Storage
Description
- Easily sealed using commercially available heat-sealers
- Binds negligible amounts of proteins or active substances
Specifications
Specifications
| Array | 16 x 24 |
| Material | Cycloolefin Copolymer |
| No. of Wells | 384 |