missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Small Self-Laminating Blank Tags

Catalog No. 19112942
Change view
Click to view available options
Color:
Red
White
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19112942 White
19112944 Red
19112943 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19112942 Supplier Brady™ Supplier No. 101973
Only null left

Prevent your text from smearing or smudging in some of the harshest environments. Brady™ Small Self-Laminating Blank Tags are durable one-sided tags with a write-on area that accepts most marking pens.

  • Polyester tags provide identification solutions for almost any environment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are water-resistant

Specifications

Quantity 25/Pk.
Color White
Material Polyester
Includes (25) nylon fasteners
Shape Rectangle
Waterproof Water-resistant
Dimensions (L x W) 1.5 x 3 in.
Label Type Self-laminating