Learn More
Eisco™ Small Air Cored Solenoid
Study electricity and magnetism using an external magnetic field. The Small Air Cored Solenoid is excellent for studying electricity and magnetism in experiments requiring an external magnetic field
Supplier:  Eisco™ PH0804D
Catalog No. S12973
Only null left
Explore available promotions
Description
Description
  • 700 turns
  • Insulated copper
  • 1.6 amps continuous current
  • 5 amps intermittent current
  • Air Cored 1.77 in. diameter, 5.9 in. length
Specifications
Specifications
Magnetism, Electromagnetism
Small Solenoid
15 cm
9 to 12
700 turns of insulated copper wire on square plaques with 4 mm sockets.
Product Suggestions
Product Suggestions