missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Pierce™ Pierce™ SM(PEG)2, No-Weigh™ Format
Description
Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging helps prevent loss of reagent reactivity and contamination over time by minimizing repetitive opening and closing of vial. The format enables use of fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.
- Reactive groups: NHS ester and maleimide
- Reactive toward: amino and sulfhydryl groups
- Irreversibly crosslink proteins or peptides by flexible PEG spacer arms
- Polyethylene glycol spacer arms help maintain conjugate solubility
- Pure compound with defined structure and molecular weight, ensuring reproducible protein-modification effects
- PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
- Ideal for performing controlled, two-step protein conjugations (see Sulfo-SMCC)
Specifications
Specifications
| Chemical Reactivity | Amine-Sulfhydryl |
| Cleavable | No |
| Description | SM(PEG)2 |
| Molecular Weight (g/mol) | 425.39 |
| PEGylated | Yes |
| Spacer Arm Length | 17.6 Å |
| Content And Storage | Upon receipt store desiccated at -20°C. |
| Cell Permeability | No |
| Shipping Condition | Wet Ice |
| Product Line | No-Weigh, Pierce |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.