Learn More
Raven Environmental Coretaker Sludge Sampler 16ft DFS Item
Supplier:  Raven Environmental Products Inc B1010416
Core sampler for measuring and viewing settled sludge and solids. CORETAKER has durable 1.25 in. OD clear polycarbonate tubes with black ABS machined Slip Lock Coupler and automatic check valve. No threaded fittings that break. Fittings are CNC machined for precision fit. Internal O-Rings prevent leaks. Thanks to a foam lanyard the entire unit will float if dropped in a tank. Comes in 8 ft. sections to minimize the number of couplers. Commonly used in wastewater treatment plants to monitor activated sludge blanket levels. Retrieve samples and spin in centrifuge to monitor Percent Solids for Waste Activated Sludge (WAS). Also used in onsite wastewater and septic systems. Packaged one unit per box. Exclusive Lifetime Break-Free Guarantee. Made in USA.
Catalog No. NC0639531
Only null left
Explore available promotions