missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SlowFade™ Gold Antifade Mountant
GREENER_CHOICE

Catalog No. p-9056001
Encompass
Change view
Click to view available options
Quantity:
2 mL
10 mL
5 x 2 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S36937 5 x 2 mL
S36940 2 mL
S36936 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S36937 Supplier Invitrogen™ Supplier No. S36937
Only null left

Protects fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments.

SlowFade Gold Antifade Mountant is a liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides. It contains chemical components designed to protect fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments. SlowFade Gold Antifade Mountant is glycerol-based, with no mixing or curing required, which makes it ideal for immediate viewing of the sample. It comes ready-to-use—just apply a drop to the sample, add a coverslip, and image. It is available with or without DAPI nuclear stain.

Key attributes:
• Protects dyes from fading during imaging
• Ready-to-use liquid, ideal for immediate sample viewing
• Mounted samples are stable for weeks
• Maintains signal strength—little to no quenching
• Ideal for Alexa Fluor dyes

SlowFade Gold Antifade Mountant is not recommended for mounting samples containing fluorescent proteins, such as GFP. For superior antifade protection of fluorescent proteins and fluorescent dyes in cultured cells or thin tissue slices, SlowFade Diamond Antifade Mountant is recommended. For high resolution or to image thicker tissue or 3D cell culture with a focal depth of 0–500 μm, try SlowFade Glass Antifade Mountant.

Specifications

Content And Storage Storage at room temperature is recommended but can also be stored frozen (-5° to -30°C). Protect from light.
For Use With (Equipment) Microscope
Product Line SlowFade
Product Type Mounting Media
Quantity 5 x 2 mL
Solution Type Anti-fade Mountant
Shipping Condition Room Temperature
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.