Learn More
3M™ Safety-Walk™ Slip Resistant General Purpose Tape, Series 600
Mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.
Supplier:  3M™ Safety-Walk™ 7000052247
 View more versions of this product
Catalog No. 29-636-1331
Only null left
Explore available promotions
Description
Description
  • Anti-slip tape for light to heavy shoe-traffic areas
  • High quality abrasive particles to ensure long-lasting friction in any application
  • Highly durable surface withstands the rigors of equipment traffic
  • High performance, pressure-sensitive adhesive for easy application
  • Custom shapes and sizes are also available
  • NFSI High Traction Certification
Specifications
Specifications
Yellow
Product Suggestions
Product Suggestions