missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ SlimLine™ Buckets with Casters

Catalog No. 19065103
Change view
Click to view available options
Color:
Dark Blue
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19065103 Yellow
19065100 Dark Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19065103 Supplier Micronova™ Supplier No. B7YEAC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SlimLine™ narrow polypropylene buckets are for use with the SlimLine™ or Slim-T™ cleanroom cart system. Available in blue, red, green or yellow.

For use with SlimLine™ Cleanroom Cart System

  • For use with SlimLine or SlimT Cleanroom Cart System
  • 6 gallon capacity
  • Autoclavable
  • Available in blue, red, green or yellow

Specifications

Product Type SlimLine Bucket
Material Polypropylene Bucket, Nickel-plated Caster, Thermoplastic Rubber Wheel
Autoclavable Autoclavable
Capacity (English) 6 gal.
Care Instructions Store at Ambient Temperature. To Autoclave - do not stack - invert Bucket to allow drainage.
Color Yellow
For Use With (Application) Cleaning Walls, Floors and Ceilings
For Use With (Equipment) SlimLine or SlimT Cleanroom Cart System
Sterility Non-sterile