missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ SlimLine™ Buckets with Casters

Catalog No. 19065100
Change view
Click to view available options
Color:
Dark Blue
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19065100 Dark Blue
19065103 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19065100 Supplier Micronova™ Supplier No. B7DBAC

SlimLine™ narrow polypropylene buckets are for use with the SlimLine™ or Slim-T™ cleanroom cart system. Available in blue, red, green or yellow.

For use with SlimLine™ Cleanroom Cart System

  • For use with SlimLine or SlimT Cleanroom Cart System
  • 6 gallon capacity
  • Autoclavable
  • Available in blue, red, green or yellow

Specifications

Product Type SlimLine Bucket
Material Polypropylene Bucket, Nickel-plated Caster, Thermoplastic Rubber Wheel
Autoclavable Autoclavable
Care Instructions Store at Ambient Temperature. To Autoclave - do not stack - invert Bucket to allow drainage.
Color Dark Blue
For Use With (Application) Cleaning Walls, Floors and Ceilings
For Use With (Equipment) SlimLine or SlimT Cleanroom Cart System
Sterility Non-sterile