missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ SlideMop™ Mop Heads, Irradiated
GREENER_CHOICE

Catalog No. 19116748
Change view
Click to view available options
Material:
Plastic, Polyester
Stainless Steel, Polyester
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19116748 Stainless Steel, Polyester
19000066 Plastic, Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19116748 Supplier Micronova™ Supplier No. MZCR6316IR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mop covers for use with the Micronova™ SlideMop™. The SlideMop is a simple, effective mop system featuring sturdy, collapsible plates that easily slide into the quilted polyester mop cover and lock into place with an effortless downward push.

  • Polyester mop covers for use with the the Micronova SlideMop™ Flat Head Mop. (For SlideMop frame, see QSM16.)
  • Quilted polyester mop covers with polyester pad interiors for disinfectant dispersalManufactured for use in Controlled Environments, ISO 5 thru 9
  • Good chemical resistance

Specifications

Product Type Irradiated Mop Slipcover
Head Type Flat
Color White
Length (English) 16 in.
Sterility Sterile
Length (Metric) 41 cm
Autoclavable Not Autoclavable
Disposable Reusable
Care Instructions Item may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
For Use With (Equipment) Stainless Steel Frame
For Use With (Application) Cleaning and Sanitization of Cleanroom Surfaces
Color-coded No
Material Stainless Steel, Polyester
Show More Show Less