Learn More
Epredia™ SlideMate™ TSP (Thermal Transfer Slide Printer) with Total Care Package
Numéro de catalogue. 22021860
Modifier l'affichage
Click to view available options
Capacité de lame:
144 Slides
Numéro de catalogue. Capacité de lame
22021860 144 Slides
1 options
Numéro de catalogue. 22021860 Fournisseur Epredia™ Code fournisseur 8000066B
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Produces durable 300 dpi permanent printing on microscope slide tabs. Includes 1-year warranty and 4 years of SWAP Service.
  • Protects specimens and slides
  • Direct-to-slide printing minimizes traceability errors associated with label transfer
  • Lateral movement reduces the risk of chipped slides
  • Enclosed hoppers protect against laboratory dust, debris, and contamination
  • Reliable performance for modern laboratory workflows
  • Compatible with a wide range of slide types
  • Thermal transfer technology supports versatile printing
  • Screen pitch adjustment enhances ergonomics
  • Compact footprint enables placement at the microtome
  • Supports on-demand printing
  • Total care package qualification requires validated slides
Specifications
Type de produit Slide Printer
Capacité de lame 144 Slides
Includes 1 year of warranty and 4 years of SWAP Service from Epredia. In order to qualify for Total Care Package, customer must use validated Epredia slides.