Learn More
Epredia™ SlideMate™ Pro Microscope Slide Printer
A smart solution with flexible configuration options to match your laboratory’s needs.
Supplier:  Epredia™ A83900015
Catalog No. A83900015
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • The SlideMate™ Pro printer features a dual-hopper loading system with intelligent slide selection for efficient performance.
  • Avoid wasted time switching between multiple slide types.
  • Maximize throughput by loading both hoppers with matching slides.
  • The SlideMate™ Pro has a high throughput: 4-5 seconds per slide.
  • Setup is simple.
  • The SlideMate™ Pro software uses a Windows™ based driver that easily connects to other computers and laboratory information systems.
  • SlideMate™ Pro enables sample-tracking solutions including user log-in information, time and date stamp on each slide, and unique transaction IDs.
Specifications
Specifications
Thermal Printer
330 mm
100/240 VAC 50/60Hz 1.2A Max.
25 x 75 x 1 mm (1 x 3 in.)
163 mm
Product Suggestions
Product Suggestions