Learn More
Epredia™ SlideMate™ Accessories
Catalog No. B81320041
Change view
Click to view available options
Product Type:
Crossover Ethernet Cable
Digital Imager
Keyboard
LabWriter Software
Mouse
Thermal Transfer Ribbon
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
B81320041 Thermal Transfer Ribbon
B81310001 LabWriter Software
B35310006 Digital Imager
B81310005 Crossover Ethernet Cable
361022466 Keyboard
361022467 Mouse
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. B81320041 Supplier Epredia™ Supplier No. B81320041
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Enhance and optimize the Epredia™ SlideMate™ Slide Printer with Epredia™ SlideMate Slide Printer Accessories.
Accessories include:
  • Collection system Collection trays (box of 4)
  • Hopper
  • Hot foil tape, box of 5 (roll of tape prints approx. 20,000 cassettes)
Specifications
Product Type Thermal Transfer Ribbon
For Use With (Equipment) SlideMate™ Slide Printer
Quantity Box of 1 (prints approx. 12,000 slides per roll of tape)