Learn More
Epredia™ SlideMate™ AS Slide Printer
Reduce specimen identification errors while enhancing lean workflow with the Epredia™ SlideMate™ AS On-Demand Slide Printer.
Supplier:  Epredia™ A83900001
 View more versions of this product
Catalog No. A83900001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The SlideMate AS promotes single piece workflow though unique features designed specifically for on-demand printing.
  • Fully integrated color touch-screen and simple software are easy to use
  • Built-in barcode scanner reads cassette information and imports the data to the slide label
  • Connects easily to other computers or Laboratory Information Systems
  • Prints directly onto the slide, eliminating the need for hand writing or labels
Specifications
Specifications
Slide Printer AS Basic
Black
1 in. x 3 in. or 25mm x 75mm x 1mm
SlideMate AS basic printer with color touch-screen, USB connection, power cord and simple software to print externally generated slide label only
230 mm
122 mm
50/60 Hz
1 Roll, approximately 12,000 Slides
Thermal transfer ribbon
5 Sec./Slide
9.1 in.
4.8 in.
1.2 A Max.
100/240 VAC
Product Suggestions
Product Suggestions