Learn More
Electron Microscopy Sciences Slide Warmer for 23 slides, 110 V
Supplier: Electron Microscopy Sciences 7131910
These slide warmers are ideal for use in cytology, histology, pathology and biology for paraffin tissue section mounting. Available in two sizes. Features thermostatic heat control, anodized black surface provides contrast, and LED temperature display, both set temperature and display temperature. Thermostat setting range from room temperature to 70 degree C.
Footprint: 10" x 7"