Learn More
Laxco™ SLi6PRO Fluorescent Upgrade Kit
This Fluorescent Upgrade Kit offers DAPI, FITC, and TRITC modules to SLi6 PRO Series Microscopes. Choose from assorted objective sets.
Supplier:  Laxco™ SLI6PFLKIT3
 View more versions of this product
Catalog No. SLI6PFLKIT3
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Choose an objective set
  • Includes protecting board
  • Excitation Filters:
    • 410 nm to 415 nm for DAPI
    • 480 nm to 485 nm for FITC
    • 550 nm to 555 nm for TRITC
  • Emission Filters
    • 455 nm ± 25 nm for DAPI
    • 525 nm ± 18 nm for FITC
    • 605 nm ± 17.5 nm for TRITC
  • Dichroic Filter
    • Triple bandpass for DAPI, FITC, TRITC
Specifications
Specifications
4X, 10X, 20X, 40X
SLi6 PRO Series Microscopes
Emission and Dichroic Filters (DAPI, FITC, TRITC), Corresponding RBG Filtersets
LED
1.7MP, Cooled, Color, CMOS
SLi6P-FL-Kit3
Educational Microscope
Plan Fluorescence
5 years optical and mechanical, 1 year electrical from date of purchase
WF 10X/22 mm
Trinocular
Computer
SLiPRO
Product Suggestions
Product Suggestions