missing translation for 'onlineSavingsMsg'
Learn More

Laxco™ SLi3 PRO Series Fluorescent Upgrade Kit
GREENER_CHOICE

Catalog No. SLI3PFLKIT1
Change view
Click to view available options
Model:
SLi3P-FL-Kit1
SLi3P-FL-Kit4
Objective Types:
Plan Fluorescence
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Objective Types
SLI3PFLKIT1 SLi3P-FL-Kit1 Plan Fluorescence
SLI3PFLKIT4 SLi3P-FL-Kit4 Plan Fluorescence
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SLI3PFLKIT1 Supplier Laxco™ Supplier No. SLI3PFLKIT1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Fluorescent Upgrade Kit offers DAPI, FITC, and TRITC modules to SLi3 PRO Series Microscopes. Choose from assorted objective sets.

  • Excitation Filters:
    • 410 nm to 415 nm for DAPI
    • 480 nm to 485 nm for FITC
    • 550 nm to 555 nm for TRITC
  • Emission Filters
    • 455 nm ± 25 nm for DAPI
    • 525 nm ± 18 nm for FITC
    • 605 nm ± 17.5 nm for TRITC
  • Dichroic Filter
    • Triple bandpass for DAPI, FITC, TRITC

Specifications

Magnification Power 4X, 10X, 20X, 40X
Objective Types Plan Fluorescence
For Use With (Equipment) SLi3 PRO Series Microscopes
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase
Filter Type Emission and Dichroic Filters (DAPI, FITC, TRITC)
Eyepiece 10X/22 mm
Lighting Type LED
Head Style Trinocular
Camera Type None
Display Type Computer
Model SLi3P-FL-Kit1
Product Type Educational Microscope
Show More Show Less