missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SLC4A4 Recombinant Protein Antigen

Numéro de catalogue. NBP232020PE
Modifier l'affichage
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
NBP232020PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. NBP232020PE Fournisseur R&D Systems™ Code fournisseur NBP232020PEP

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human SLC4A4. The SLC4A4 Recombinant Protein Antigen is derived from E. coli. The SLC4A4 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP2-32020. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

Spécifications

Gene ID (Entrez) 8671
Species Human
Product Line Novus
Purification Method Chromatography
Purity >80%
Concentration 0.5mg/mL
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Symbol SLC4A4
Label Type Unlabeled
Molecular Weight (g/mol) 24kDa
Product Type SLC4A4
Quantity 0.1 mL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-32020. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogen KKKGSLDSDNDDSDCPYSEKVPSIKIPMDIMEQQPFLSDSKPSDRERSPTFLERHTS
Afficher plus de résultats Afficher moins de résultats

For Research Use Only