missing translation for 'onlineSavingsMsg'
Learn More

Carson™ SkySeeker™ Newtonian Reflector Telescope with Dobsonian Mount, 15x - 37.5x

Catalog No. S98152
Encompass
Change view
Click to view available options
Magnification:
15 to 37.5x
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification
S98152 15 to 37.5x
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98152 Supplier Carson™ Supplier No. EPDJC200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Features precision 76 mm optics and a power range of 15x to 37.5x for high-quality imaging, includes 8 mm and 20 mm Huygenian eyepieces.

  • Precision 76 mm optics and a 15x to 37.5x power range for high-quality imaging
  • Provides amazing images of the moon and planets, as well as views of bright deep sky objects like the Orion Nebula (M42), Hercules Cluster (M13) and Andromeda Galaxy (M31)
  • Includes 8 mm and 20 mm Huygenian eyepieces
  • Ready to use right out of the box, no assembly required
  • One year limited warranty

Specifications

Model JC-200
Mount Dobsonian
Warranty 1 Year, Limited
Product Type Reflector Telescope
Aperature 76 mm
Includes 8 mm and 20 mm Huygenian Eyepieces, Dobsonian Mount
Weight (English) 3.7 lb.
Magnification 15 to 37.5x