Learn More
Ergodyne™ Skullerz™ Dagr Safety Glasses/Sunglasses, Black Frame
Protective work safety sunglasses feature tough scratch-resistant lenses that filter 99.9% of harmful UVA, UVB and UVC rays to keep eyes safe in the sun and the blade-style design is a sleek and sporty look.
$27.51 - $60.92
Specifications
| Lens Coating | Scratch-resistant, UV-treated |
|---|---|
| Certifications/Compliance | ANSI Z87.1+, CSA Z94.3 + and MIL-PRF 32432 Ballistic Impact |
| For Use With (Application) | Construction/Trades, Oil and Gas, Mining, Maintenance, Assembly/Fabrication, Manufacturing, Police/Tactical |
| Frame Color | Black |
| No. per Case | 12/Cs. |
| Catalog Number | Mfr. No. | Lens Tint | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Lens Tint | Price | Quantity | |||||
|
19-910-288
|
Ergodyne™
52000 |
Clear |
Each for $27.51
|
|
|||||
|
19-910-289
|
Ergodyne™
52003 |
Clear with Fog-Off™ |
Each for $32.40
|
|
|||||
|
19-910-293
|
Ergodyne™
52031 |
Polarized Smoke |
Each for $60.92
|
|
|||||
|
19-910-294
|
Ergodyne™
52033 |
Smoke with Fog-Off™ |
Each for $32.40
|
|
|||||
|
19-910-295
|
Ergodyne™
52042 |
Silver, Mirror |
Each for $27.51
|
|
|||||
|
19-910-296
|
Ergodyne™
52050 |
Yellow |
Each for $27.51
|
|
|||||
|
19-910-297
|
Ergodyne™
52071 |
Polarized G15 |
Each for $60.92
|
|
|||||
|
19-910-298
|
Ergodyne™
52080 |
Indoor/Outdoor |
Each for $27.51
|
|
|||||
|
19-910-299
|
Ergodyne™
52083 |
Indoor/Outdoor with Fog-Off™ |
Each for $32.40
|
|
|||||
|
19-910-300
|
Ergodyne™
52092 |
Blue, Mirror |
Each for $27.51
|
|
|||||
Description
- Durable half frames bend and flex without breaking
- Non-slip rubber nose piece and rubber temples keep the glasses secure and resist sweat
- Available with a polarized lens or with anti-fogging solution
- Polarized glasses reduce glare and eye fatigue around water, snow and in other bright environments.
- Anti-glare glasses block the harsh reflected light from water
- Fog-Off™ treatment won't wash off or wear off for fog-free viewing and are ideal for use with dust masks
- Certifications and testing:
- Impact-resistant safety glasses are tested to ANSI Z87.1+ High-Velocity Impact Protection and optical requirements
- Tested to MIL-PRF-32432 Ballistic Fragmentation, Class 1, which is the US Military Combat Eye Protection System Performance Specification for eyewear. To pass the military standard, safety glasses must withstand a steel projectile shot at 660 ft / 201m per second which is 4x faster than the Z87+ standard. Lenses must not puncture, crack or shatter. Frames must retain the lenses and not break.
- Ideal for outdoor work like construction, carpentry/woodworking or landscaping or for outdoor activities like boating, skiing, fishing, hunting/shooting, and sports.
- Available in a variety of lens options ideally suited for a variety of tasks and light environments
Specifications
| Scratch-resistant, UV-treated | |
| Construction/Trades, Oil and Gas, Mining, Maintenance, Assembly/Fabrication, Manufacturing, Police/Tactical | |
| 12/Cs. |
| ANSI Z87.1+, CSA Z94.3 + and MIL-PRF 32432 Ballistic Impact | |
| Black |