Learn More
Pyramex™ Sitecore™ Safety Eyewear
Hard coated polycarbonate, scratch-resistant lens
$68.00 - $68.00
Specifications
| Certifications/Compliance | ANSI Z87.1,CSA Z94.3 |
|---|---|
| Certifications/Compliance (Europe) | EN-166 |
| Weight (Metric) | 24.4 g |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |
| Catalog Number | Mfr. No. | Frame Color | Lens Tint | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Frame Color | Lens Tint | Price | Quantity | ||||
|
19-155-890
|
Pyramex™
SBG10110D |
Black, Gray | Clear |
Pack of 12 Each for $68.00
|
|
||||
Description
Specifications
| ANSI Z87.1,CSA Z94.3 | |
| 24.4 g | |
| Polycarbonate |
| EN-166 | |
| Polycarbonate |