missing translation for 'onlineSavingsMsg'
Learn More

Sisomicin Sulfate 98.0+%, TCI America™

Catalog No. p-7131727
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I1049250MG 250 mg
I10491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I1049250MG Supplier TCI America Supplier No. I1049250MG
Only null left

Chemical Identifiers

CAS 53179-09-2
Molecular Formula C19H39N5O11S
Molecular Weight (g/mol) 545.605
MDL Number MFCD00058308
InChI Key JVTNJDPXUPRGIE-DAVQCCERSA-N
Synonym sisomicin sulfate
PubChem CID 123134107
IUPAC Name (2S,3R,4S,5S)-2-[(2S,3S,4R,6S)-4,6-diamino-3-[[3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid
SMILES CC1(COC(C(C1NC)O)OC2C(CC(C(C2O)OC3C(CC=C(O3)CN)N)N)N)O.OS(=O)(=O)O

Specifications

Melting Point 73°C
Color White
Boiling Point 155°C
Quantity 250 mg
Formula Weight 692.71
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sisomicin Sulfate
Recommended Storage Refrigerator