Learn More
Mopec Single Scrub Station
Description
- Thermostatically-controlled mixing faucet with gooseneck spout
- Backflow protection for all plumbing fixtures to prevent backflow of contaminated water
- Rapid and positive drainage
- Access panel to get to plumbing
- ADA wheelchair accessible: (SS101-ADA)
- OA-151: Hot and cold foot valve
Specifications
Specifications
| Certifications/Compliance | Non - ADA |
| Color | Silver |
| Material | Stainless Steel (304) |
| Type | Hand Sink |