Learn More
Micro Essential Lab Single-Roll Hydrion™ pH Test Paper, With Chart
For measuring pH solutions.
Supplier:  Micro Essential Lab 51
 Afficher plus de versions de ce produit
Numéro de catalogue. MES51
Il en reste null
Description
Description
  • Used in laboratories, hospitals and universities worldwide
  • Comes with a 7/32 in. x 15 ft. roll of test paper and matching color chart
  • Provides for approximately 100 tests
Spécifications
Identifiants chimiques
Colorimetric chart
100 Tests
pH Test Paper
Spécifications
1 to 11
Colorimetric chart
pH Test Paper
For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing
100 Tests
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS