Learn More
Micro Essential Lab Single-Roll Hydrion™ pH Test Paper, With Chart
For measuring pH solutions.
Supplier: Micro Essential Lab 55
Description
- Used in laboratories, hospitals and universities worldwide
- Comes with a 7/32 in. x 15 ft. roll of test paper and matching color chart
- Provides for approximately 100 tests
Chemical Identifiers
| Colorimetric chart | |
| 100 Tests | |
| pH Test Paper |
Specifications
| 4 to 9 | |
| Colorimetric chart | |
| pH Test Paper |
| For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing | |
| 100 Tests |