Learn More
Micro Essential Lab Single-Roll Hydrion™ pH Test Paper, With Chart
For measuring pH solutions.
Supplier:  Micro Essential Lab 52
 View more versions of this product
Catalog No. 14853150P
Only null left
Description
Description
  • Used in laboratories, hospitals and universities worldwide
  • Comes with a 7/32 in. x 15 ft. roll of test paper and matching color chart
  • Provides for approximately 100 tests
Specifications
Chemical Identifiers
Colorimetric chart
100 Tests
pH Test Paper
Specifications
1 to 6
Colorimetric chart
pH Test Paper
For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing
100 Tests
Product Suggestions
Product Suggestions