Learn More
Micro Essential Lab Single-Roll Hydrion™ pH Test Paper, With Chart
For measuring pH solutions.
Supplier: Micro Essential Lab #375
Description
- Used in laboratories, hospitals and universities worldwide
- Comes with a 7/32 in. x 15 ft. roll of test paper and matching color chart
- Provides for approximately 100 tests
Chemical Identifiers
| Colorimetric chart | |
| 100 Tests | |
| pH Test Paper |
Specifications
| 10 to 12 | |
| Colorimetric chart | |
| pH Test Paper |
| For laboratories, hospitals and universities worldwide | |
| 100 Tests |