Learn More
Ihc World Llc SINGLE EDGE RAZOR BLADE 100/PK
Catalog No. NC0305367
Encompass
Top quality & most popular single edge blades made w/ high carbon surgical steel. Precision honed. Each blade is individually wrapped. Ideal for cutting & trimming tissue specimen, paraffin & frozen blocks, as well as other general laboratory usage.
Catalog No. NC0305367 Supplier Ihc World Llc Supplier No. IW2100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Top quality & most popular single edge blades made w/ high carbon surgical steel. Precision honed. Each blade is individually wrapped. Ideal for cutting & trimming tissue specimen, paraffin & frozen blocks, as well as other general laboratory usage.