missing translation for 'onlineSavingsMsg'
Learn More

Simvastatin 97.0+%, TCI America™

Catalog No. S0509100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0509100MG 100 mg
S05091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0509100MG Supplier TCI America Supplier No. S0509100MG

Chemical Identifiers

CAS 79902-63-9
Molecular Formula C25H38O5
Molecular Weight (g/mol) 418.574
MDL Number MFCD00072007
InChI Key RYMZZMVNJRMUDD-HGQWONQESA-N
Synonym simvastatin, zocor, synvinolin, sinvacor, denan, lipex, simvastatine, sivastin, lodales, simvastatina
PubChem CID 54454
ChEBI CHEBI:9150
IUPAC Name [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbutanoate
SMILES CCC(C)(C)C(=O)OC1CC(C=C2C1C(C(C=C2)C)CCC3CC(CC(=O)O3)O)C

Specifications

Melting Point 134°C
Color White
Quantity 100 mg
Formula Weight 418.57
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Simvastatin
RTECSNumber EK7798000
TSCA No