missing translation for 'onlineSavingsMsg'
Learn More

Simvastatin, 98%

CAS: 79902-63-9 | C25H38O5 | 418.57 g/mol

Supplier:  Thermo Scientific Chemicals 458840010

 View more versions of this product

Catalog No. AC458840010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Simvastatin
127.0°C to 132.0°C
Authentic
Glass Bottle
1 g
RYMZZMVNJRMUDD-HGQWONQESA-N
[(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbutanoate
54454
418.57
79902-63-9
0.5% max.
97.5% min. (HPLC)
C25H38O5
simvastatin, zocor, synvinolin, sinvacor, denan, lipex, simvastatine, sivastin, lodales, simvastatina
CCC(C)(C)C(=O)OC1CC(C=C2C1C(C(C=C2)C)CCC3CC(CC(=O)O3)O)C
418.57
CHEBI:9150
98%

Chemical Identifiers

79902-63-9
418.57
simvastatin, zocor, synvinolin, sinvacor, denan, lipex, simvastatine, sivastin, lodales, simvastatina
CHEBI:9150
CCC(C)(C)C(=O)OC1CC(C=C2C1C(C(C=C2)C)CCC3CC(CC(=O)O3)O)C
C25H38O5
RYMZZMVNJRMUDD-HGQWONQESA-N
54454
[(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbutanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
Obtain special instructions before use.
IF exposed or concerned:

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only