missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Urine Collection System: Urine Tubes Only

Catalog No. 22046100
Change view
Click to view available options
Volume (Metric):
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22046100 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22046100 Supplier Simport™ Scientific Supplier No. T4101*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Urine Tubes for use with T410 Simport Urine Collection System.

  • Urine tubes are made of virgin polystyrene and are free of any mold release agents, metals or additives that could contaminate samples.
  • They are made of heavy wall construction, graduated at 1/4, 1/2, 1, 2 and every 2 mL thereafter up to 12 mL, and can be safely centrifuged at speeds up to 2000 g.
  • The tubes are flared at the top to make filling and drip-free pouring easier.
  • Designed to allow the use of midget urinometers and reagent test strips requiring only 1/4 or 1/2 mL of sample.
  • Size of tube: 105 mm H x 21 mm dia.

Specifications

Product Type Urine Tubes
Disposable Single-use
Material Polystyrene
Diameter (Metric) Inner 21 mm
For Use With (Equipment) Urine Collection System, 36-101-5251 (T410)
Graduations 0.25 to 12 mL
Height (English) 4.1 in.
Height (Metric) 105 mm
Includes 500 Disposable 15 mL heavy-wall Polystyrene tubes
No. per Pack 500 Pk.
Volume (English) 0.5 oz.
Volume (Metric) 15 mL
Show More Show Less