Learn More
Simport™ Scientific Swingsette™ Tissue Cassettes In Quickload™ Sleeves
Suitable for hoppers accepting plastic sleeves, these Swingsette™ Tissue Cassettes In Quickload™ Sleeves will load in cassette labeling instruments in one simple operation. Just load the 75-cassette QuickLoad™ sleeve in the hopper and you are ready for printing.
Supplier: Simport™ Scientific M51711SL
Description
- Made of acetal
- Sleeved cassettes for use with Thermo Fisher printers
- Anterior writing area at a 45° angle
- Large tab for convenient and easy opening
- Covers packaged separately
- Easy-to-assemble disposable cover with special hinge
Specifications
| Orange | |
| Tissue Cassette |