Learn More
Simport™ Scientific Swingsette™ Tissue Cassettes In Quickload™ Sleeves
Catalog No. 361019535
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
361019535 White
361019536 Pink
22038643 Peach
22038644 Tan
361019539 Blue
22038645 Gray
22038647 Aqua
361019541 Orange
361019538 Yellow
361019540 Lilac
361019537 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 361019535 Supplier Simport™ Scientific Supplier No. M5172SL
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Suitable for hoppers accepting plastic sleeves, these Swingsette™ Tissue Cassettes In Quickload™ Sleeves will load in cassette labeling instruments in one simple operation. Just load the 75-cassette QuickLoad™ sleeve in the hopper and you are ready for printing.
  • Made of acetal
  • Sleeved cassettes for use with Thermo Fisher printers
  • Anterior writing area at a 45° angle
  • Large tab for convenient and easy opening
  • Covers packaged separately
  • Easy-to-assemble disposable cover with special hinge
Specifications
Color White
Product Type Tissue Cassette