Learn More
Simport™ Scientific Swingsette™ Tissue Cassettes In Quickload™ Sleeves
Suitable for hoppers accepting plastic sleeves, these Swingsette™ Tissue Cassettes In Quickload™ Sleeves will load in cassette labeling instruments in one simple operation. Just load the 75-cassette QuickLoad™ sleeve in the hopper and you are ready for printing.
Supplier:  Simport™ Scientific M5177SL
 View more versions of this product
Catalog No. 22-038-643
Only null left
Explore available promotions
Description
Description
  • Made of acetal
  • Sleeved cassettes for use with Thermo Fisher printers
  • Anterior writing area at a 45° angle
  • Large tab for convenient and easy opening
  • Covers packaged separately
  • Easy-to-assemble disposable cover with special hinge
Specifications
Specifications
Peach
Tissue Cassette
Product Suggestions
Product Suggestions