Learn More
Simport™ Scientific Swingsette™ Biopsy Processing/Embedding Cassettes
Catalog No. 22038704
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Closure Type:
Covers Packaged Separately
Removable Cover
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type
22038704 Aqua Covers Packaged Separately
22038670 Pink Removable Cover
22038669 White Removable Cover
22038672 Yellow Removable Cover
22038673 Blue Removable Cover
22038674 Peach Removable Cover
22038676 Tan Removable Cover
22038679 Orange Removable Cover
22038677 Gray Removable Cover
22038680 Aqua Removable Cover
22038678 Lilac Removable Cover
361012035 White Covers Packaged Separately
22038694 Green Covers Packaged Separately
22038693 Pink Covers Packaged Separately
22038695 Yellow Covers Packaged Separately
22038697 Peach Covers Packaged Separately
22038696 Blue Covers Packaged Separately
22038698 Tan Covers Packaged Separately
22038699 Gray Covers Packaged Separately
22038702 Lilac Covers Packaged Separately
22038703 Orange Covers Packaged Separately
22038671 Green Removable Cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Catalog No. 22038704 Supplier Simport™ Scientific Supplier No. M51812
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These Swingsette™ Biopsy Cassettes are specially designed to hold biopsy specimens during the processing/embedding process as well as in storage cabinets. Molded from acetal, they keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents.
  • Made of acetal
  • Anterior writing area at a 45° angle
  • Large tab for convenient and easy opening
  • Easy-to-remove pre-attached cover with special hinge
  • Can be opened and closed as often as necessary
  • 1 mm square openings to maximize fluid exchange
Specifications
Packaging Type Covers Packaged Separately
Closure Type Covers Packaged Separately
Color Aqua
Material High Density Polymer
Product Type Biopsy Cassette