Learn More
Simport™ Scientific Swingsette™ Biopsy Cassettes In Quickload™ Sleeves
Suitable for hoppers accepting plastic sleeves, these Swingsette™ Biopsy Cassettes In Quickload™ Sleeves will load in cassette labeling instruments in one simple operation. Specially designed to hold biopsy specimens during the processing/embedding process as well as in storage cabinets.
Supplier: Simport™ Scientific M5185SL
Description
- Made of acetal
- 1 mm square openings to maximize fluid exchange
- Sleeved cassettes for use with ThermoFisher printers
- Anterior writing area at a 45° angle
- Large tab for convenient and easy opening
- Covers packaged separately
- Easy-to-assemble disposable cover with special hinge
Specifications
| Yellow | |
| Biopsy Cassette |