missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ Scientific Tamper Evident Self-Standing Transport Tubes
Description
- For secure short and long term storage
- Provide strength and clarity
- Non-toxic, non sterile polypropylene made etched or non graduated tubes
- Exhibit unique design features
- Unique transport tubes incorporate a seal that needs to be broken in order to open it, which leaves visual indication that the vial has been opened
- With external threads for providing smooth and uniform inner surface
- Closure offers unique frangible band design
- Combination of tamper evident and resistant closure system
- For clinical trials and expensive reagents in diagnostic kits
- Caps sold separately
Specifications
Specifications
| Product Type | Transport Tube (Self-Standing) |
| Diameter (Metric) | 16.6 mm |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 5 mL |
| Bottom Shape | Conical |
| Material | Polypropylene |
| For Use With (Application) | Storage and transportation of biological materials |
| Length (Metric) | 65 mm |
| Sterility | Non-sterile |
| Graduated | Yes |
| Show More |