Learn More
Simport™ Scientific SLIMSETTE™ M510SL Biopsy Processing/Embedding Cassettes in Quickload™ Sleeves
SLIMSETTE™ M510SL Biopsy Processing/Embedding Cassettes in Quickload™ Sleeves are a new generation of cassettes, prepackaged for use in automatic cassette labeling instruments.
Supplier: Simport™ Scientific M5106SL
Description
- Specially designed to hold biopsy specimens during the embedding process
- Made of acetal polymer
- Transparent sleeve allows clear view of cassettes during printing
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- Lid can be opened and closed as often as necessary, and will always relock securely
- Large labeling area for easy identification
- Anterior writing area at a 45° angle
Chemical Identifiers
| Slots | |
| 45° | |
| Acetal Polymer | |
| Biopsy Cassette |
Specifications
| Slots | |
| Integral Lid | |
| Acetal Polymer |
| 45° | |
| Blue | |
| Biopsy Cassette |