Learn More
Simport™ Scientific SlideFile™ Storage System
The SlideFile™ Storage System is a convenient, organized, and versatile way of storing 75 x 25 mm or 3 x 1 in. microscope slides.
Supplier: Simport™ Scientific M700100P
Description
- Holds up to 400 microscope slides per unit (with slots removed)
- Stores 75 x 25 mm or 3 x 1 in. microscope slides
- 82 W x 245 L x 86 mm H
- 1720 cm3 (105 cu. in.)
- Impact resistant
- Stackable
- Tinted, hinged cover
- Base available in different colors to help in slide classification
- All slides stored upright for easier insertion and removal
- Allows reading of bar codes without having to remove the slides from the box
- Not autoclavable
Specifications
| 400 | |
| 9.64 in. | |
| 400 | |
| 86 mm | |
| 3.22 in. |
| Pink | |
| 245 mm | |
| 3.38 in. | |
| Slide box, removable tray | |
| 82 mm |