Learn More
Simport™ Scientific OneHand™ Microtube Rack
The OneHand™ Microtube Rack is compact, lightweight and stackable in order to save as much space as possible and can be used all around the lab. This is why it is ideal for incubators, refrigerators, freezers, under lab hoods and on bench tops.
Supplier:  Simport™ Scientific T36050O
 View more versions of this product
Catalog No. 05-409-10
Only null left
Explore available promotions
Description
Description
  • Made of acetal
  • Locking base hold self standing Micrewtube™ in place for opening and closing
  • Submersible but maintains stability without tipping over
  • Interlocking feet allow for safe stacking
Specifications
Specifications
Microtube Rack
Orange
Acetal Polymer
10/Cs.
11.5 in.
4.5 in.
1.5 in.
Not Autoclavable
Microtubes (0.5 to 2 mL)
50
29.3 cm
11.45 cm
3.9 cm
Product Suggestions
Product Suggestions